Learn More
ACROBiosystems Biotinylated Human HLA-C*07:02:01 & B2M & RYR ,His,Avitag™&Tag Free (HLR-H52W2) is expressed from human 293 cells (HEK293). It contains AA Cys 25 – Ile 308 | Ile 21 - Met 119 (Accession # P10321-1 | RYRPGTVAL & P61769-1).
Supplier:  ACROBiosystems HLRH52W225UG
Encompass_Preferred
HLA-A, B, and C are transmembrane glycoproteins in the major histocompatibility complex 1 (MHC I) family. The C receptor is a heterodimer consisting of a HLA-C mature gene product (heavy chain) and β2-microglobulin (light chain). The mature C chain is anchored in the membrane. HLA class I molecules play a central role in the immune system by presenting peptides derived from endoplasmic reticulum lumen. HLA-C are expressed in nearly all cells, and present small peptides to the immune system which surveys for non-self peptides.
Catalog No. 50-225-3430
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.