Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AACS/ACSF1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981319
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1319 Supplier Bioss Supplier No. BS11383R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Activates acetoacetate to acetoacetyl-CoA. May be involved in utilizing ketone body for the fatty acid-synthesis during adipose tissue development.

Specifications

Antigen AACS/ACSF1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AACS
Gene Accession No. Q86V21
Gene Alias 2210408B16Rik; aacs; AcAc-CoA ligase; acetoacetate-CoA ligase; acetoacetyl-CoA ligase; Acetoacetyl-CoA synthetase; ACSF1; Acyl-CoA synthetase family member 1; BOS_16499; homolog of C. elegans supressor of ras 5 (sur-5); Protein sur-5 homolog; si:dkey-215k6.2; SUR5; SUR-5; zgc:56105
Gene Symbols AACS
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AACS .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65985
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.