Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human AAK1, GST Tag Recombinant Protein

Catalog No. A33370
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A33370 1 mg
A30967 10 μg
A32880 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A33370 Supplier Invitrogen™ Supplier No. A33370
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This AAK1 recombinant human protein (1-465) was expressed in insect cells. The purified catalytic domain (1-510) of AAK1 can be used in a number of different applications, but has been specifically validated for use with our LanthaScreen™ Kinase binding assay technology to identify novel kinase inhibitors. AAK1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Catalytic.

AAK1 serine/threonine kinase is a member of the NAK family and plays a regulatory role in endocytosis. AAK1 is a member of the SNF1 subfamily of Ser/Thr protein kinases. The protein interacts with and phosphorylates a subunit of the AP-2 complex, which promotes binding of AP-2 to sorting signals found in membrane-bound receptors and subsequent receptor endocytosis. Its kinase activity is stimulated by clathrin.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q2M2I8
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 0.25mM DTT, 0.1mM PMSF, 150mM NaCl, 0.1mM EDTA, 10mM glutathione and no preservative; pH 7.5
Gene ID (Entrez) 22848
Molecular Weight (g/mol) 77.3 kDa
Name Human AAK1, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 5530400K14Rik; 9630042K20; Aak1; Adaptor-associated kinase 1; AP2 associated kinase 1; AP2-associated protein kinase 1; AU067724; AU067726; BC028270; C79663; D6Ertd245e; DKFZp686F03202; DKFZp686K16132; FLJ23712; FLJ25931; FLJ31060; FLJ42882; FLJ45252; KIAA1048; MGC138170; MGC164568; MGC164570; mKIAA1048; R75501; RGD1563580
Common Name AAK1
Gene Symbol AAK1
Kinase Family NAK Kinase Family
Protein Length 1-510
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-510
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.