Learn More
Medchemexpress LLC 6-(3,4-dichlorobenzamido)-N-(thiazol-2-yl)-2-naphthamide | 2247919-28-2 | 98.1% | 442.32 g/mol | C21H13Cl2N3O2S | 5MG
SDP
Supplier:  Medchemexpress LLC HY1262495MG
Encompass_Preferred
AAPK-25 is a potent, selective dual inhibitor of Aurora kinases and Polo-like kinases (PLK) used in preclinical research to investigate cell cycle regulation and anticancer mechanisms. It causes mitotic delay and arrests cells in prometaphase, and is provided as a solid or as a DMSO solution for cellular and biochemical assays.
  • Potent dual inhibitor of Aurora kinases and PLK.
  • Induces mitotic delay and prometaphase arrest in cells.
  • Suitable for cellular and biochemical kinase assays.
  • High purity (98.1%) for research applications.
  • Available in solid and solution formats for flexible use.
  • Simple storage guidance for sample stability in solution and solid forms.
Catalog No. 50-002-39641
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.