Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AAR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578461 Supplier Invitrogen™ Supplier No. PA578461
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: IMR32 Predicted Reactivity: Mouse (88%), Rat (88%), Bovine (91%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

AAR2 encodes the homolog of the yeast A1-alpha2 repressin protein that is involved in mRNA splicing. Alternately spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AAR2
Applications Western Blot
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene AAR2
Gene Accession No. Q9Y312
Gene Alias 0610011L14Rik; Aar2; AAR2 splicing factor homolog; AAR2 splicing factor homolog (S. cerevisiae); C20orf4; CGI-23; novel transcript; PRO0225; Protein AAR2 homolog; RGD1311066
Gene Symbols AAR2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AAR2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25980
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.