Learn More
American Research Products Inc AAV-6 TITRATION ELISA
Supplier:  American Research Products Inc PRAAV6
Encompass
Unique microtiter plate enzyme immunoassay for the quantitation of intact AAV6 wt virions, AAV6 recombinant virions or assembled and intact empty capsids of human Adeno-associated Virus 6. The capture-antibody detects a conformational epitope not present on unassembled capsid proteins.
Catalog No. NC1745512
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.