Learn More
Teknova AAV Affinity CIP2 Solution, 1000mL, Non-Sterile
SDP
Supplier:  Teknova 20771
Encompass_Preferred
6M Guanidine Hydrochloride
Catalog No. 50-238-7782
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.