Learn More
Teknova AAV Affinity Storage Buffer, 1000mL, Sterile 3/UN1170/II
SDP
Supplier:  Teknova 20772
Encompass
39mM Sodium Phosphate Monobasic, 61mM Sodium Phosphate Dibasic, 20% Ethanol, adjusted to pH 7.0
Catalog No. 50-238-7819
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.