Learn More
Teknova AAV•Tek™ AEX Buffer Screening Kit: AAV8
SDP
Supplier:  Teknova 20780
Encompass
Through extensive screening, we created a first-of-its-kind kit that can help you identify the ideal buffer formulation for the separation of empty and full capsids while maintaining high yields, saving months in your process development at the polishing step. Each serotype-specific kit contains paired sets of optimized equilibration and elution buffers in 1 L bottles. These discrete formulations have been proven to be effective across multiple upstream and purification platforms. For AAV8.
Catalog No. 50-238-7731
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.