Learn More
ACROBiosystems AAV3 Titration ELISA Kit
Supplier:  ACROBiosystems AAVA00396TESTS
Encompass_Preferred
AAV3 Titration ELISA Kit is based on ELISA sandwich method and designed to detect and quantitative determine of AAV3 capsid titration in AAV gene therapy product preparation processing. It contains AAV3 empty capsids and a pair of antibodies against AAV3. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of AAV3 capsid titration.
Catalog No. 50-253-5533
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.