Learn More
System Biosciences LLC AAVS1 SparQ(TM) All-In-One Inducible System, 1 Kit
SDP
Supplier:  System Biosciences LLC GEQM820AKIT
Encompass
GEQM820A-1(AAVS1-SA-RFP-Puro-EF1-CymR-CuO-MCS-P2A-GFP) with CAS601A-1 (All-in-one Cas9 SmartNuclease AAVS1 Targeting Plasmid) and GEQM840PR-1 (Junction PCR Primer Mix to confirm AAVS1 site integration)
Catalog No. NC2738195