Learn More
Medchemexpress LLC HY-129393 5mg Medchemexpress, AB928 CAS:2239273-34-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1293935MG
Encompass_Preferred
Medchemexpress, HY-129393 5mg AB928 CAS:2239273-34-6 AB928 is an orally bioavailable, selective dual adenosine receptor (A2aR/A2bR) antagonist with immunomodulatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9806
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.