Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Abacavir hemisulfate

Catalog No. 414810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-481-0 10 mg
41-485-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-481-0 Supplier Tocris Bioscience™ Supplier No. 4148/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Reverse transcriptase inhibitor; antiretroviral

Abacavir hemisulfate is a competitive reverse transcriptase inhibitor; inhibits HIV replication. Orally active and brain penetrant. Antiretroviral agent.

Specifications

Purity 99%
Molecular Formula (1 S,4 R)-4-[2-Amino-6-(cyclopropylamino)-9 H-purin-9-yl]-2-cyclopentene-1-methanol hemisulfate
CAS 188062-50-2
Content And Storage 4°C
Quantity 10 mg
Product Type Abacavir hemisulfate
Molecular Weight (g/mol) M.W. (Observed): 335.37
Inhibitors Abacavir hemisulfate

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.