Learn More
ALLIED ANALYTIC REFURB HM5C AND VS2 ABAXIS VETSCAN HM5C HEMATOLOGY AND VS2 CHEM ANALYZER COMBO,
Supplier:  ALLIED ANALYTIC 790000012001001
Encompass_Preferred
*This is a refurbished item. Certified like-new. The VetScan HM5 is a fully-automated, five-part differential hematology analyzer displaying a comprehensive 22-parameter complete blood count (CBC) with cellular histograms on an easy-to-read touch-screen. Its superior performance, elegant design, ease of use, true database management capability, and minimal maintenance make it the optimal hematology system for veterinary clinics, research laboratories, and pharmaceutical and biotech companies.
Catalog No. NC2171877
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.