Learn More
Anton Paar Digital Performance Refractometer – Abbemat 300
Supplier:  Anton Paar 97790
Encompass_Preferred
Abbemat 300 refractometer allows non-destructive refractive index and concentration measurements within seconds. Independent of sample properties you can measure liquids, pastes, polymers, solids, turbid, colored, or opaque samples. - Routine quality control measurements of refractive index and concentration at a precision of +/-0.0001 nD- In-built application knowhow with over 200 methods for your sample- Fully 21CFR part 11 compliance- Avoids human errors by showing a clear pass/fail result- Connects to other Anton Paar instruments for multiparameter measurementsMeasuring range Refractive Index scale (RI) - Range 1.26 to 1.72 nD- Resolution ± 0.00001 nD- Accuracy ± 0.0001 nDMeasuring range Brix scale- Range 0 to 100 % - Resolution 0.01 % - Accuracy 0.05 % Temperature- Range 4 to 85 °C- Accuracy ± 0.05 °C- Stability ± 0.002 °CAccessories: NC1818555, NC1818545, NC1818543
Catalog No. NC1818534
Only null left
Add to cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.