Learn More
Anton Paar Digital Performance Plus Refractometer – Abbemat 550
Supplier:  Anton Paar 97787
Encompass_Preferred
Abbemat 550 refractometers allow non-destructive refractive index and concentration measurements within seconds. Independent of sample properties you can measure liquids, pastes, polymers, solids, turbid, colored, or opaque samples. - Highest precision (+/-0.00002 nD), ideal for R&D and advanced quality control applications- In-built application knowhow with over 200 methods for your sample- Full 21 CFR Part 11 compliance with complete pharma feature set and advanced data integrity software solutions - Avoids human errors by showing a clear pass/fail result- Connects to other Anton Paar instruments for multiparameter measurementsMeasuring range Refractive index scale (RI) - Range nD 1.26 to 1.72 - Resolution nD ± 0.000001 - Accuracy nD ± 0.00002 Measuring range Brix scale- Range 0 to 100 % - Resolution 0.01 % - Accuracy 0.05 % Temperature- Range 4 to 85 °C- Accuracy ± 0.03 °C - Stability ± 0.002 °CAccessories: NC1818555, NC1818545, NC1818543, NC1818551
Catalog No. NC1818539
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.