Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ AbC™ Anti-Rat/Hamster Bead Kit

Catalog No. A10389
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10389 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10389 Supplier Molecular Probes™ Supplier No. A10389
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

AbC™ Anti-Rat/Hamster Bead Kit

The AbC™ Anti-Rat/Hamster Antibody Compensation Bead Kit provides a consistent, accurate, and simple-to-use technique for the setting of flow cytometry compensation when using fluorochrome-conjugated to rat or hamster antibodies with any laser in flow cytometry.

  • Species reactivity—for use with all isotypes of rat and hamster antibody conjugates
  • Maximum laser compatibility—for use with all lasers

    Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Often, however, cellular-baseThe AbC™ Anti-Mouse Antibody Compensation Bead Kit provides a consistent, accurate, and simple-to-use technique for the setting of flow cytometry compensation when using fluorochrome-conjugated mouse antibodies with any laser in flow cytometry.

  • Species reactivity—for use with all isotypes of mouse antibody conjugates
  • Maximum laser compatibility—for use with all lasers

    Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Often, however, cellular-based compensation controls are not completely effective, as many antigens are not highly expressed, and dimly stained cells can lead to inaccurate compensation settings. The AbC™ Anti-Mouse Bead Kit was developed to address this issue.

    How it works
    Each kit provides polystyrene microspheres that have fluorescent antibody conjugate capture capacity (positive compensation beads) or are inert (negative compensation beads). After mixing with a fluorophore-conjugated mouse antibody, the two components provide a distinct high-signal positive control with an appropriate negative population that can then be used to set compensation properly regardless of the intensity of the cells in the actual experiment.

    Species reactivity
    The AbC™ Anti-Mouse Bead Kit is ideal for compensation with all isotypes of mouse (IgG1, IgG2a, IgG2b, IgG3, IgM) immunoglobulins.

    Maximum laser compatibility
    All AbC™ compensation bead kits are ideal for use with any excitation source, including the 405 nm laser which has historically provided higher background signals due to auto-fluorescence with other kits on the market.d compensation controls are not completely effective, as many antigens are not highly expressed, and dimly stained cells can lead to inaccurate compensation settings. The AbC™ Anti-Rat/Hamster Bead Kit was developed to address this issue.

    How it works
    Each kit provides polystyrene microspheres that have fluorescent antibody conjugate capture capacity (positive compensation beads) or are inert (negative compensation beads). After mixing with a fluorophore-conjugated rat or hamster antibody, the two components provide a distinct high-signal positive control with an appropriate negative population that can then be used to set compensation properly regardless of the intensity of the cells in the actual experiment.

    Species reactivity
    The AbC™ Anti-Rat/Hamster Bead Kit is ideal for compensation with all isotypes of rat (IgG1, IgG2a, IgG2b, IgG2c, IgM) and hamster (Armenian IgG and Syrian IgG) immunoglobulins.

    Maximum laser compatibility
    All AbC™ Compensation Bead Kits are ideal for use with any excitation source, including the 405 nm laser which has historically provided higher background signals due to auto-fluorescence with other kits on the market.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Diameter (Metric) 6 μm
    Content And Storage Contains 1 vial AbC™ anti-rat/hamster capture beads and 1 vial of negative beads.
  • Store at 2°C to 8°C
  • Emission any lasers (UV to 633/635)
    Format Tube
    For Use With (Equipment) Flow Cytometer
    Quantity 1 kit
    Detection Method Fluorescence
    Stain Configuration Surface
    Shipping Condition Room Temperature
    Product Line CompenFlow, Molecular Probes
    Product Type Compensation Beads
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.