Learn More
ABC Vial™, Sold by MilliporeSigma™ Supelco™
Catalog No. 111009205 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Color:
Amber
Clear
Marking Spot:
No
Yes
No. per Pack:
100
1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Marking Spot No. per Pack
11-100-9205 Amber Yes 1000
11-100-9305 Clear Yes 1000
11-100-9591 Clear No 1000
11-101-1787 Amber Yes 100
11-101-1915 Clear No 100
11-101-1876 Clear Yes 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 11-100-9205 Supplier MilliporeSigma Supelco Supplier No. 27557
Add to Cart
Add to Cart
Use with autosamplers that require 12 x 32 mm screw top or crimp top vials. 40% wider opening than standard screw cap vials. Superior thread design ensures a secure seal. Compatible with most GC autosamplers that accept crimp top. Can be used with robotic systems. VialsCaps and septa are not included.
Autosampler compatibility:
  • Agilent
  • Agilent 1050/1090A
  • Agilent 1100/1200
  • Agilent 7673A, 7683
  • Agilent 5890/7985A/6890
  • Hitachi
  • AS-1000, AS-2000, AS-4000L7200/L7250
  • LEAP Technologies
  • CTC LC PAL, GC PAL
  • CombiPAL
  • HTC PAL/HTS PAL
  • PerkinElmer
  • Autosystem/AS-2000/XL
  • Clarus 500/600
  • ISS-100, 200
  • Shimadzu LC Autosamplers
  • SIL-HTa/SIL-HTc
  • SIL-10ADVP
  • SIL-10A/SIL-10Ai/SIL-10Ap
  • Varian
  • CP-8410
  • 8100/8200
  • 9100/90950/9095
  • Waters
  • Alliance 2690
  • Acquity
  • Breeze
  • 717Plus
TRUSTED_SUSTAINABILITY
Specifications
Color Amber
Material Glass
No. per Pack 1000
Volume (English) 2 mL
Marking Spot Yes
ABC Vial is a trademark of Wheaton Industries, Inc.