Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104402
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104402 Supplier Invitrogen™ Supplier No. PA5104402
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Abca6.

The ATP-binding cassette (ABC) superfamily is comprised of transmembrane proteins involved in energy-dependent transport of a variety of substrates across membranes. ABCA6 is a 1,617 amino acid protein belonging to the ABC transporter family. ABCA6 is up-regulated during monocyte differentiation into macrophages, suggesting a role in macrophage lipid homeostasis. ABCA6 contains two ABC transporter domains and is expressed as three isoforms produced by alternative splicing. ABCA6 is present at highest levels in adult liver, with significant levels found in many adult tissues and fetal lung, kidney and liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA6
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ABCA6
Gene Accession No. Q8K441, Q8N139
Gene Alias 6330565N06Rik; ABC transporter ABCA6; Abca6; ATP binding cassette subfamily A member 6; ATP-binding cassette A6; ATP-binding cassette sub-family A member 6; ATP-binding cassette transporter sub-family A member 6; ATP-binding cassette, subfamily A (ABC1), member 6; ATP-binding cassette, sub-family A (ABC1), member 6; EST155051
Gene Symbols ABCA6
Host Species Rabbit
Immunogen A synthesized peptide derived from mouse Abca6(Accession Q8K441), corresponding to amino acid residues L1589-A1609.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23460, 303639, 76184
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.