Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559624
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559624 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559624 Supplier Invitrogen™ Supplier No. PA559624
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VNRTFEELTL LRDVREVWEM LGPRIFTFMN DSSNVAMLQR LLQMQDEGRR QPRPGGRDHM EALRSFLDPG SGGYSWQDAH ADVGHL Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 60%.

ATP-binding cassette (ABC) transporters are an evolutionarily conserved family that use ATP hydrolysis to catalyze the transport of various molecules across cell membranes. They are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). ABCA7, a member of the ABC1 subfamily, is a full-size ABC transporter consisting of two sets of the multiple membrane-spanning domains plus the Walker motifs for the ATP interaction. ABCA7 shows the highest homology to ABCA1, an essential molecule for cholesterol homeostasis. The high expression levels of ABCA7 in peripheral leukocytes, thymus, spleen and bone marrow suggests a role in the immune system lipid homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Abca7
Gene Accession No. Q8IZY2
Gene Alias ABC 1 antibody; ABC Transporter 1; ABC1; ABC1 antibody; Abc51; Abca7; ABCA-SSN; ABCX; AD9; ATP binding cassette subfamily A member 7; ATP-binding cassette sub-family A member 7; ATP-binding cassette sub-family A member 7; LOW QUALITY PROTEIN: ATP-binding cassette sub-family A member 7; ATP-binding cassette, subfamily A (ABC1), member 7; ATP-binding cassette, sub-family A (ABC1), member 7; ATP-binding cassette, sub-family A, member 7; autoantigen SS-N; CERP; CERP antibody; EC 2.2.1.1; EC 3.6.3; EC 3.6.3.41; FLJ40025; Macrophage ABC transporter; Phospholipid-transporting ATPase ABCA7; phospholipid-transporting ATPase ABCA7; ATP-binding cassette sub-family A member 7
Gene Symbols Abca7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABCA7. Recombinant protein control fragment (Product #RP-98889).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10347
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.