Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Abcam, 100X NAD+ Assay Kit, 1 Unit

Catalog No. 502672428
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-267-2428 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-267-2428 Supplier Abcam Supplier No. AB2522751UNIT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly concentrated reagent for use in NAD+ detection assays.

100X NAD+ is a reagent provided in assay kit format for research use.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule NAD+
Concentration 100X
Substrate Properties Chemical Substrate
Substrate NAD+
Quantity 1 Unit
For Use With (Application) Assay kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.