Learn More
Abcam alpha-L-Fucosidase Assay Kit, 100 Tests
Description
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Conjugate | Unconjugated |
| Gene Symbols | FUCA1 |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Primary |
| Packaging | Vial |
| Target Species | Non-species specific |
| Product Type | Antibody |
| Detection Method | Colorimetric |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.