Learn More
Abcam Abcam, DNMT Activity Assay Kit, Fluorometric, 96 Tests
Description
- Assay Type: Enzyme activity
- Detection Method: Fluorescent
- Assay Time: 3 hr 45 min
- Applications: Enzyme activity assay
- Cited in: 1 publication
- Recommended Equipment: Microplate reader
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | DNMT |
| For Use With (Equipment) | Microplate Reader |
| No. of Reactions | 96 Tests |
| Run Time | 3 hr 45 min. |
| Detection Method | Fluorescence |
| Format | Kit |
| Quantity | 96 Tests |
| For Use With (Application) | Enzyme activity assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.