Learn More
Abcam Human IL-18 Matched Antibody Pair Kit, 5 x 96 Tests
Description
- Applications: ELISA
- Species Reactivity: Human
Specifications
Specifications
| Applications | ELISA |
| Conjugate | Unconjugated |
| Gene Symbols | IL18 |
| Quantity | 5 x 96 Tests |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Test Specificity | This product reacts with Human. |
| Target Species | Human |
| Product Type | Secondary Antibody |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.