Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Human/Monkey TNF alpha Antibody Pair - BSA and Azide free, 10 x 96 Tests

Catalog No. 502663426
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-266-3426 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-266-3426 Supplier Abcam Supplier No. AB24179110X96TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody, suitable for sELISA.

  • Applications: sELISA
  • Species Reactivity: Human, Rhesus monkey and Cynomolgus monkey

Specifications

Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols TNF
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Human, Rhesus monkey and Cynomolgus monkey.
Target Species Human, Rhesus Monkey, Cynomolgus Monkey
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Hu, RhMk
Validated Application abrv sELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.