Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Human TNF alpha Matched Antibody Pair Kit, 5 x 96 Tests

Catalog No. 502588355
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-8355 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-8355 Supplier Abcam Supplier No. AB2134675X96TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody, suitable for ELISA, IA, with 2 citations.

  • Applications: ELISA, IA
  • Species Reactivity: Human

Specifications

Applications ELISA
Conjugate Unconjugated
Gene Symbols TNF
Quantity 5 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Human.
Target Species Human
Product Type Secondary Antibody
Detection Method Colorimetric
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv ELISA, IA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.