Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Mouse Alanine aminotransferase (ALT) Antibody Pair - BSA and Azide free, 10 x 96 Tests

Catalog No. 502663257
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-266-3257 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-266-3257 Supplier Abcam Supplier No. AB25364710X96TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody, suitable for sELISA.

  • Applications: sELISA
  • Species Reactivity: Mouse

Specifications

Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols Gpt
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv sELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.