Learn More
Abcam Mouse Hemoglobin subunit alpha Antibody Pair - BSA and Azide free, 10 x 96 Tests
Description
- Applications: sELISA
- Species Reactivity: Mouse
Specifications
Specifications
| Applications | Sandwich ELISA |
| Conjugate | Unconjugated |
| Gene Symbols | Hba |
| Quantity | 10 x 96 Tests |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Test Specificity | This product reacts with Mouse. |
| Target Species | Mouse |
| Product Type | Secondary Antibody |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.