Learn More
Abcam Abcam, NADP Regeneration Kit, 200 Tests
Description
The NADP Regeneration Kit is designed for applications requiring NADP regeneration.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Format | Kit |
| No. of Reactions | 200 Tests |
| Substrate Properties | Chemical Substrate |
| Substrate | NADP+ |
| Target Molecule | NADP |
| Quantity | 200 Tests |
| For Use With (Application) | Functional assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.