Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572939 Supplier Invitrogen™ Supplier No. PA572939
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABCG1 Polyclonal Antibody for Western Blot, IHC (P), Flow

ATP-binding cassette (ABC) transporter genes are involved in the regulation of the amount of dietary cholesterol retained in the body. The ABC transporter ABCG1 (ABC8) is expressed on the cell surface and in intracellular compartments of cholesterol-laden macrophages and is induced in monocyte-derived macrophages during cholesterol influx mediated by acetylated low-density lipoprotein. In contrast, lipid efflux in cholesterol-laden macrophages, mediated by the cholesterol acceptor HDL(3), suppresses the expression of ABCG1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCG1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ABCG1
Gene Accession No. P45844, Q64343
Gene Alias ABC transporter 8; ABC8; ABCG1; ATP binding cassette subfamily G member 1; ATP-binding cassette 8; ATP-binding cassette sub-family G member 1; ATP-binding cassette transporter 8; ATP-binding cassette transporter member 1 of subfamily G; ATP-binding cassette, subfamily G (WHITE), member 1; ATP-binding cassette, sub-family G (WHITE), member 1; ATP-binding cassette, subfamily G, member 1; AW413978; homolog of Drosophila white; I79_011524; MGC34313; White; White protein homolog; white protein homolog (ATP-binding cassette transporter 8); WHITE1; WHT1
Gene Symbols ABCG1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of human ABCG1 (between residues 300-400).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100759619, 103346773, 11307, 85264, 9619
Target Species Human, Mouse, Rat, Rabbit, Monkey, Hamster
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.