Learn More
Medchemexpress LLC Abciximab 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP99345MG
Encompass_Preferred
Abciximab (C7E3) a chimeric mouse/human monoclonal antibody fragment is a glycoprotein (GP) IIb/IIIa inhibitor Abciximab inhibits platelet aggregation and leucocyte adhesion by binding to the glycoprotein IIb/IIIa vitronectin and Mac-1 receptors[1]
Catalog No. 50-003-22581