Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam ABD-F, Fluorogenic Reagent, Thiol Labeling, 10 mg

Catalog No. 502705347
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5347 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5347 Supplier Abcam Supplier No. AB14527310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Highly reactive and sensitive fluorogenic reagent for the selective labeling of protein cysteine residues.

  • Molecular weight: 217.18 g/mol
  • Purity: >98%
  • Form: Solid
  • Composition: 100% 7-fluoro-2,1,3-benzoxadiazole-4-sulfonamide
  • Storage: Store at -20°C
  • Shipping: Ambient - Can Ship with Ice
  • Functional attributes: Highly reactive, sensitive fluorogenic reagent for thiol labelling; selectively labels protein cysteine residues without side chain modification or N-terminal blockage; contains a benzofurazan moiety that produces a highly fluorescent compound through the reaction with a sulfhydryl group

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Product Type Protein Labeling Reagent
Content And Storage Store at -20°C.
Form Solid
Target Molecule Thiol
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrate Sulfhydryl group
Detection Method Fluorescence
Formulation 7-fluoro-2,1,3-benzoxadiazole-4-sulfonamide
Quantity 10 mg
For Use With (Application) Thiol labelling
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.