Learn More
Antistat Inc. Abeba Uni 6 Slip on Shoe
Description
- Made from microfiber, these shoes are specifically designed for all day comfort
- Breathable antibacterial lining
- Energy absorbing heel area
- Soft PU midsole for additional cushioning
- Water and chemical repellent
- ESD dissipative: conforms to DIN EN 61340
- Washable to 30°C
Specifications
Specifications
| Color | White |
| Material | Microfiber |
| Size | 46 |
| Waterproof | Yes |
| Product Type | Cleanroom Shoes |