Learn More
TARGETMOL CHEMICALS INC AB-680 5MG
Supplier: TARGETMOL CHEMICALS INC T10216
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AB-680 is a highly potent reversible and selective inhibitor of CD73 (an ectonucleotidase Ki 4.9 pM for hCD73) with anti-tumor activity displays >10000-fold selectivity over related ectonucleotidases CD39. purity: 97%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.