Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Abgene™ 2D Barcoded 5mL Screw Cap Tubes
GREENER_CHOICE

Catalog No. AB1470 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
AB1470 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1470 Supplier Thermo Scientific™ Supplier No. AB1470
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Abgene 2D Barcoded 5 mL Screw Cap tubes, ideal for storing larger volumes, are encapsulated with laser-etched 2D barcode on the base to securely identify and track samples. Our technical experts are available to demonstrate ways to improve sample management in your laboratory.

The Abgene 2D Barcoded 5mL Screw Cap tubes, ideal for storing larger volumes, are encapsulated with laser-etched 2D barcode on the base to securely identify and track samples. Our technical experts are available to demonstrate ways to improve sample management in your laboratory. We offer a full range of Thermo Scientific™ 2D barcoded storage tubes, racks and readers as well as pipettes and automated liquid handling equipment.

Secure, Fast Tracking

  • Scan the tube into any application or database, or integrate via various data export options
  • Instantly access sample information and identify contents by scanning the tube using the database Find feature
  • Barcodes can be instantly decoded by Thermo Scientific barcode readers

Efficient Storage, Superior Design

  • Save precious space with the dense microplate footprint (48 tubes per rack) of stackable Thermo Scientific Abgene freezer racks
  • Grid-referenced lids enable you to easily see the storage tube location
  • External threaded tubes aid in contamination prevention

Stringent Quality Control

  • Every storage tube code is scanned to guarantee readability
  • Each code is checked against the entire database of all 2D barcoded storage tubes to ensure zero duplicates
  • Every lot of storage tubes is leak tested to ensure the integrity and security of samples
  • All tubes are certified free of DNase, RNase and DNA

Recommended for:

Individual sample retrieval; Instant sample identification; Secure identification of archived samples; Secure shipping of samples

TRUSTED_SUSTAINABILITY

Specifications

Description Tube with Cap, Racked
Capacity (Metric) 5 mL
Material Polypropylene
Packaging Racked
Barcode 2D BarCoded
Closure Included Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.