Learn More
Thermo Scientific™ ABgene™ 384-Well, Full Skirted, Extra Volume PCR Plates
Catalog No. BC0937
Change view
Click to view available options
Color:
Natural
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
BC0937 Natural
BC0937W White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BC0937 Supplier Thermo Scientific™ Supplier No. BC0937
Thermo Scientific™ ABgene 384-Well Full Skirted Extra Volume PCR Plates have increased well volume making the plates suitable for use in post-PCR applications.
Thermo Scientific™ ABgene™ 384-Well, Full Skirted, Extra Volume PCR Plates feature increased well volume for convenience in post-PCR applications.
Highlights
  • Tall Chimney well design
  • Increased well volume accommodates sequencing and wash steps
  • Working well volume: 40 µL
  • Cut corner: A24
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certified free of human DNA, RNase, DNase, and endotoxins.
Format Standard
Description Full Skirted, with Barcode
Array 16 x 24
Color Natural
Material Virgin Polypropylene
No. of Wells 384
Volume (Metric) Working 40 μL