Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101396 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101396 Supplier Invitrogen™ Supplier No. PA5101396
  • $573.00
    $478.65 / Each of 1

    Save $94.35

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ABHD6.

ABHD6 has 2-arachidonoylglycerol hydrolase activity. ABHD6 may be a regulator of endocannabinoid signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ABHD6
Gene Accession No. Q5XI64, Q8R2Y0, Q9BV23
Gene Alias 0610041D24Rik; 2-arachidonoylglycerol hydrolase; AA673485; Abhd6; abhydrolase domain containing 6; abhydrolase domain containing 6, acylglycerol lipase; abhydrolase domain-containing protein 6; AV065425; lipase protein; monoacylglycerol lipase ABHD6
Gene Symbols ABHD6
Host Species Rabbit
Immunogen A synthesized peptide derived from human ABHD6(Accession Q9BV23), corresponding to amino acid residues R236-A286.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305795, 57406, 66082
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.