Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD7 Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5145426
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145426 20 μL
FEPA5145425 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145426 Supplier Invitrogen™ Supplier No. PA5145426
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABHD7 gene ontology (GO) annotation includes hydrolase activity and membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene EPHX4
Gene Accession No. Q6IE26, Q8IUS5
Gene Alias ABHD7; abhydrolase domain containing 7; abhydrolase domain-containing protein 7; EH4; EPHX4; Ephxrp; epoxide hydrolase 4; Epoxide hydrolase-related protein; Gm1382
Gene Symbols EPHX4
Host Species Rabbit
Immunogen Recombinant Mouse EPHX4 protein, Lys36-Asp359 (Accession #Q6IE26).
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253152, 384214
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.