Learn More
Applied Biosystems™ ABI PRISM™ 7000 Sequence Detection Systems Spectral Calibration Kit
Description
Specifications
Specifications
| Content And Storage | Contains one Background plate, and seven Spectral Calibration plates with seven dye standards (FAM™, JOE™, NED™,ROX™, SYBR™ Green, VIC™, TAMRA™). Store at -15 to 25°C. Protect from light. |
| Calibrated Dye | FAM, JOE, NED, ROX, TAMRA™, TMR (Tetramethylrhodamine), VIC, SYBR™ Green |
| For Use With (Equipment) | 7000 System |
| Detection Method | Fluorescence |
| Product Line | ABI PRISM |
| Product Type | Spectral Calibration Kit |
| For Use With (Application) | Real Time PCR (qPCR) |
| Shipping Condition | Dry Ice |
| Quantity | 8 x 96-well plates |
| Calibration Type | Spectral Calibration |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.