Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566491 Supplier Invitrogen™ Supplier No. PA566491
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSSTAPDAAAG GAQTLADGFT SPTPPVVSST PPTGHPVQFY SMNRPASRHT Highest antigen sequence identity to the following orthologs - mouse 94%, rat 51%.

ABI2 may act in regulation of cell growth and transformation by interacting with nonreceptor tyrosine kinases ABL1 and/or ABL2. ABI2 is a part of the WAVE complex that regulates lamellipodia formation. The WAVE complex regulates actin filament reorganization via its interaction with the Arp2/3 complex. It regulates ABL1/c-Abl-mediated phosphorylation of MENA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABI2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Abi2
Gene Accession No. Q9NYB9
Gene Alias 8430425M24Rik; Abelson interactor 2; Abi2; abi-2; ABI2B; abl binding protein 3; abl interactor 2; Abl-binding protein 3; AblBP3; abl-interacting protein 1 (SH3-containing protein); abl-interactor 2; abl-interactor protein 2b; AI839867; AIP-1; arg protein tyrosine kinase-binding protein; arg-binding protein 1; ArgBP1; ARGBPIA; argBPIB; C130078H13; SSH3BP2; thyroid hormone responsive protein
Gene Symbols Abi2
Host Species Rabbit
Immunogen Recombinant Human ABI2. Recombinant protein control fragment (Product #RP-104252).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10152
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.