Learn More
Apexbio Technology LLC Dexrazoxane HCl (ICRF-187, ADR-529) 149003-01-0 100mg
SDP
Supplier:  Apexbio Technology LLC A1602100
Encompass_Preferred
Dexrazoxane HCl (ICRF-187 ADR-529) is a small-molecule inhibitor targeting topoisomerase II It is designed to inhibit topoisomerase II activity and sequester intracellular iron thereby reducing DNA strand cleavage and mitigating iron-dependent oxidative stress Dexrazoxane HCl exerts its biological activity primarily through intracellular iron chelation and inhibition of topoisomerase II which attenuates reactive oxygen species formation and stabilizes DNA structure Based on these pharmacological properties Dexrazoxane HCl holds research potential in investigating cardioprotective strategies particularly against anthracycline-induced cardiotoxicity
Catalog No. 50-000-94924
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.