Learn More
Eurofins Discoverx Abl (M351T), active, 250 µg. (14-757M)
Supplier:  Eurofins Discoverx 14757M
Encompass_Preferred
N-terminal 6xHis-tagged recombinant, human Abl, amino acids 27-end containing the M351T mutation. The Abl tyrosine kinase inhibitor STI571 is an effective therapy for stable-phase chronic myeloid leukemia patients. Many patients responding to STI571 later relapse due to a reactivation of Bcr-Abl activity. In certain cases this appears to correlate with the presence of the M351T mutation, which confers resistance to drug binding (Shah et al., (2002), Cancer Cell 2, 117-125). For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9296
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.