Learn More
Agilent Technologies ABLE C COMPETENT CELLS
Supplier:  Agilent Technologies 200171
Encompass
ABLE C Competent Cells 5 x 0.2mL aliquots Reduces the number of common cloning vectors 4-fold Clone DNA encoding toxic proteins Reduce plasmid copy number to increase cell viability Decrease levels of cloned protein product
Catalog No. 50-125-027
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.