Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 20S Proteasome Assay Kit

Catalog No. 89024634 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-634 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-634 Supplier Abnova™ Supplier No. KA1352
Add to Cart
Add to Cart

Used for assaying the activity of 20S proteasome and functional study

Specifications

Content And Storage Store the kit at -80°C temporarily. For best results, store the components as described in the protocol.
Description 20S Proteasome Assay Kit is a fluorescence-based method for assaying the activity of 20S proteasome.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 1 vial 20S proteasome positive control, 1 vial 20S proteasome substrate (SUC-LLVY-AMC), 1 vial epigallocatechin gallate (EGCG), 1 vial 20S proteasome assay buffer, 1 vial 20S proteasome lysis buffer, 1 plate 96-well black plate with lid
Quantity 1 Kit
Type Activity Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.