Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
7-AAD Cell Viability Assay Kit is a fluorescence-labeled method for screening dead or damaged cells.
Flow Cytometry
Specifications
Specifications
| Content And Storage | 4°C |
| Detection Method | Fluorometric |
| For Use With (Application) | Used for screening dead or damaged cells and flow cytometry |
| Includes | 50μL 7-AAD staining stock solution 1000X, 50mL 7-AAD assay fixative/Actinomycin D solution, 4 tablets buffer tablet |
| Quantity | 100 Tests |
| Type | Cell Viability Assay Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?