Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 8-Epi-PGF2-alpha ELISA Kit

Catalog No. 89100262 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-262 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-262 Supplier Abnova™ Supplier No. KA0443
Add to Cart
Add to Cart

Includes

  • 96-well micortiter plate, pre-coated with 8-epi-prostaglandin-F2α antibody, 1
  • 8-epi-prostaglandin-F2α Standards (1μg/mL), 2 x 60μL
  • Enhanced Dilution Buffer, 100mL
  • Wash Buffer (5x), 40mL
  • TMB Substrate (Tetramethylbenzidine), 25mL
  • HRP Conjugate, 250μL
  • Disposable reagent troughs for a multichannel pipettor, 2
  • Stop Solution, 5mL

Use to measure quantitatively 8-Epi-Prostaglandin F2-alpha. Sandwich enzyme immunoassay.

This 8-Epi-PGF2-alpha ELISA Kit is a competitive enzyme-linked immunoassay (ELISA) for determining levels of 8-epi-prostaglandin-F2α in urine samples

Specifications

Calibration Range 0.05 to 100ng/mL
Conjugate HRP
Description 8-Epi-Prostaglandin F2-alpha ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of 8-Epi-Prostaglandin F2-alpha.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Urine
Sample Volume 200 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type 8-Epi-PGF2-alpha ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.