Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Acetylcholinesterase Assay Kit

Catalog No. 89028339 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-339 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-339 Supplier Abnova™ Supplier No. KA1607
Add to Cart
Add to Cart

Includes

  • Assay buffer: (pH 7.5), 30mL
  • Reagent, 240mg
  • Calibrator: (equivalent to 200U/L), 4mL

Rapid colorimetric determination of acetylcholinesterase activity in blood, cell lysate, tissue lysate

Specifications

Calibration Range 10 to 600U/L
Content And Storage Store the kit at room temperature.
Description Acetylcholinesterase Assay Kit is a rapid colorimetric determination of acetylcholinesterase activity.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 30mL assay buffer, 240mg reagent, 4mL calibrator
Quantity 1 Kit
Sample Type Blood, Cell Lysate, Tissue Lysate
Sample Volume 10 μL
Type Activity Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.