Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Adipolysis Assay Kit

Catalog No. 89028391 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-391 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-391 Supplier Abnova™ Supplier No. KA1659
Add to Cart
Add to Cart

For quantitative colorimetric/fluorometer assay for adipolysis in cell culture medium

Specifications

Calibration Range 10 to 1000μM
Content And Storage Store Assay Buffer at 4°C. Store other reagents at -20°C.
Description Adipolysis Assay Kit is a quantitative colorimetric/fluorimetric assay for adipolysis.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes 24mL assay buffer, 500μL enzyme mix, 250μL ATP, 220μL dye reagent, 100μL 100mM Glycerol standard
Quantity 1 Kit
Sample Type Cell Culture Medium
Sample Volume 10 μL
Type Adipolysis Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.