Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ADIPOQ (Human) ELISA Kit

Catalog No. 89100013 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Milk, Plasma, Saliva, Serum, Urine
CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine
Plasma (Citrate, EDTA, Heparin), Serum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-013 Plasma (Citrate, EDTA, Heparin), Serum
89-028-041 CCS, Milk, Plasma, Saliva, Serum, Urine
89-100-726 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-100-012 CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 89-100-013 Supplier Abnova™ Supplier No. KA0016
Add to Cart
Add to Cart

For detection of ADIPOQ in Human Urine and Murine, Adiponectin in Plasma, Serum and cell culture supernatants

Specifications

Calibration Range NA
Conjugate HRP
Cross Reactivity Human, Monkey
Description ADIPOQ (Human) ELISA Kit is a competitive enzyme-linked immunosorbent assay for the quantitative measurement of human adiponectin in serum and plasma.
Detection Limit 26ng/mL
Detection Method Colorimetric
Includes 96 wells Antigen coated microtiter strips, Conjugate solution (7mL), Set of standards (7 x 0.22mL), Quality control HIGH (0.4mL), Quality control LOW (0.4mL), Dilution buffer (2 x 13mL), Wash solution conc. 10X (100mL), Substrate solution (2 x 13mL), Stop solution (9mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Plasma (Citrate, EDTA, Heparin), Serum
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ADIPOQ (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.