Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Adipoq (Mouse) ELISA Kit

Catalog No. 89100011 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma, Serum, Urine
CCS, Serum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-011 CCS, Serum
89-028-165 CCS, Plasma, Serum, Urine
89-100-727 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-100-011 Supplier Abnova™ Supplier No. KA0018
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Adipoq in Urine, Plasma, Serum and cell culture supernatants

Specifications

Calibration Range 0.25 to 8ng/mL
Conjugate HRP
Cross Reactivity Mouse
Description Adipoq (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Adiponectin/Acrp30 protein in serum and tissue culture medium.
Detection Limit 0.079ng/mL
Detection Method Colorimetric
Includes 96 wells Antigen coated microtiter strips, Conjugate solution (13mL), Master standards (2vials), Quality control HIGH (2vials), Quality control LOW (2vials), Dilution buffer (22mL), Wash solution conc. 10X (100mL), Substrate solution (13mL), Stop solution (13mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ADIPOQ (Mouse) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.